Abnova
Manufacturer Code:H00000159K
Catalog # 89-208-356
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ADSS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ADSS peptide using ARM Technology.
For Research Use Only
Protein Aliases: ADEH MGC20404; adenylosuccinate synthetase (Ade(-)H-complementing); Adenylosuccinate synthetase isozyme 2; Adenylosuccinate synthetase, acidic isozyme; Adenylosuccinate synthetase, liver isozyme; adSS 2; AMPSase 2; IMP--aspartate ligase 2; L-type adenylosuccinate synthetase
Gene Aliases: ADEH; ADSS; ADSS 2; ADSS2
UniProt ID: (Human) P30520
Entrez Gene ID: (Human) 159
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.