Invitrogen
Manufacturer Code:PA578826
Catalog # PIPA578826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence of human Adenylosuccinate Lyase. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: argininosuccinase; Argininosuccinate lyase; Argininosuccinate lyase ASAL Arginosuccinase ASL; Arginosuccinase; ASAL
Gene Aliases: 2510006M18Rik; ASAL; ASL
UniProt ID: (Human) P04424, (Mouse) Q91YI0, (Rat) P20673
Entrez Gene ID: (Human) 435, (Mouse) 109900, (Rat) 59085
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.