Invitrogen
Manufacturer Code:PA544973
Catalog # PIPA544973
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ADRM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 110 kDa cell membrane glycoprotein; Adhesion regulating molecule 1 ARM1 GP110 MGC29536 Rpn13; Adhesion-regulating molecule 1; ARM-1; cell membrane glycoprotein, 110000M(r) (surface antigen); glycoprotein 110; Gp110; hRpn13; M(r) 110,000 surface antigen; Proteasomal ubiquitin receptor ADRM1; Proteasome regulatory particle non-ATPase 13; Rpn13 homolog
Gene Aliases: 1110063P18Rik; 2510006J17Rik; AA408205; ADRM1; ARM-1; ARM1; AU043535; GP110; Rpn13
UniProt ID: (Human) Q16186, (Rat) Q9JMB5, (Mouse) Q9JKV1
Entrez Gene ID: (Human) 11047, (Rat) 65138, (Guinea pig) 100736167, (Mouse) 56436
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.