Invitrogen
Manufacturer Code:PA5109364
Catalog # PIPA5109364
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ADRA2B (NP_000673.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [a]2B ADA2B Adra2b Adra-2b ADRA2L1 ADRA2RL1 ADRARL1 Alpha-2 adrenergic receptor subtype C2 alpha2B Alpha-2B adrenergic receptor Alpha-2B adrenoceptor Alpha-2B adrenoreceptor ALPHA2BAR alpha-2BAR alpha2-C2 FAME2; ADRA2B adrenergic, alpha-2B-, receptor; Adrenergic alpha2B- receptor class III; adrenergic receptor, alpha 2b; adrenergic, alpha-2B-, receptor; Adrenergic, alpha2B-, receptor class III; Alpha-2 adrenergic receptor subtype C2; alpha-2-adrenergic receptor-like 1; Alpha-2B adrenergic receptor; Alpha-2B adrenoceptor; Alpha-2B adrenoreceptor; alpha-2BAR; G-protein coupled receptor; subtype alpha2-C2
Gene Aliases: [a]2B; Adra-2b; ADRA2B; ADRA2L1; ADRA2RL1; ADRARL1; alpha-2BAR; alpha2-C2; alpha2B; ALPHA2BAR; FAME2
UniProt ID: (Human) P18089, (Rat) P19328
Entrez Gene ID: (Human) 151, (Mouse) 11552, (Rat) 24174
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.