OriGene
Manufacturer Code:TA890050
Catalog # 50-168-4763
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human ADRA2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADRA2 ADRA2R ADRAR ALPHA2AAR ZNF32; adrenergic, alpha-2A-, receptor; Alpha-2 adrenergic receptor subtype C10; alpha-2-adrenergic receptor, platelet type; Alpha-2A adrenergic receptor; Alpha-2A adrenoceptor; Alpha-2A adrenoreceptor; alpha-2AAR; alpha-2AAR subtype C10; alpha2A-adrenergic receptor; subtype alpha2-C10
Gene Aliases: Adra-2; Adra-2a; ADRA2; ADRA2A; ADRA2R; ADRAR; alpha(2A)AR; alpha2-C10; alpha2A; alpha2A-AR; ALPHA2AAR; AW122659; ZNF32
UniProt ID: (Human) P08913, (Mouse) Q01338
Entrez Gene ID: (Human) 150, (Mouse) 11551
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.