Abnova
Manufacturer Code:H00113622B01P
Catalog # 89-933-521
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ADPRHL1 (NP_6124391 a.a. &bsim 354 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human ADPRHL1 protein.
For Research Use Only
Protein Aliases: [Protein ADP-ribosylarginine] hydrolase-like protein 1; ADP-ribosyl-hydrolase; ADP-ribosylarginine hydrolase like 1; ADP-ribosylhydrolase 2; ARH2
Gene Aliases: ADPRHL1; ARH2
UniProt ID: (Human) Q8NDY3
Entrez Gene ID: (Human) 113622
Molecular Function: G-protein modulator enzyme modulator hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.