Invitrogen
Manufacturer Code:PA5114826
Catalog # PIPA5114826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ADPGK corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610017G09Rik ADP dependent glucokinase ADP-dependent glucokinase ADPGK ADP-GK ATP-dependent glucokinase C77577 EC 2.7.1.147 PSEC0260 RbBP-35 RGD1306103; ADP-dependent glucokinase; ADP-GK; ATP-dependent glucokinase; RbBP-35
Gene Aliases: 2610017G09Rik; ADP-GK; ADPGK; C77577; PSEC0260; RGD1306103
UniProt ID: (Human) Q9BRR6, (Mouse) Q8VDL4
Entrez Gene ID: (Human) 83440, (Mouse) 72141, (Rat) 315722
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.