Abnova
Manufacturer Code:H00083440A01
Catalog # 89-107-401
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 425 a.a. to 496 a.a) partial recombinant protein with GST tag.,ADPGK (NP_112574 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant ADPGK.
For Research Use Only
Protein Aliases: 2610017G09Rik; ADP-dependent glucokinase; ADP-GK; ATP-dependent glucokinase; DKFZp434B195; RbBP-35
Gene Aliases: 2610017G09Rik; ADP-GK; ADPGK; PSEC0260
UniProt ID: (Human) Q9BRR6
Entrez Gene ID: (Human) 83440
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.