Abnova
Manufacturer Code:H00000419B01
Catalog # 89-004-942
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ART3 (NP_001170.2 1 a.a. ~ 378 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human ART3 protein.
For Research Use Only
Protein Aliases: ADP-ribosyltransferase C2 and C3 toxin-like 3; ARTC3; Ecto-ADP-ribosyltransferase 3; FLJ26404; Mono(ADP-ribosyl)transferase 3; mono-ADP-ribosyltransferase; NAD(P)(+)--arginine ADP-ribosyltransferase 3
Gene Aliases: ART3; ARTC3; TMART
UniProt ID: (Human) Q13508
Entrez Gene ID: (Human) 419
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.