Novus Biologicals
Manufacturer Code:NBP297042AF405
Catalog # NB041722
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant ADP-ribosylarginine hydrolase (NP_001116.1 Met1-Leu351). |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [Protein ADP-ribosylarginine] hydrolase; [Protein ADP-ribosylarginine] hydrolase ADP-ribosylarginine hydrolaseARH1ADP-ribose-L-arginine cleaving enzyme EC 3.2.2.19; ADP-ribose-L-arginine cleaving enzyme; ADP-ribosylarginine hydrolase
Gene Aliases: ADPRH; ARH1
UniProt ID: (Human) P54922
Entrez Gene ID: (Human) 141
Molecular Function: G-protein modulator enzyme modulator hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.