Novus Biologicals
Manufacturer Code:NBP299496C
Catalog # NB085387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human ADP-Sugar Pyrophosphatase/NUDT5. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 8-oxo-dGDP phosphatase; ADP-sugar pyrophosphatase; EC 3.6.1.- EC 3.6.1.13 hYSAH1 nucleoside diphosphate linked moiety X-type motif 5 Nucleoside diphosphate-linked moiety X motif 5 nudix (nucleoside diphosphate linked moiety X)-type motif 5 Nudix motif 5 YSA1 YSA1HADP-sugar pyrophosphatase; Nuclear ATP-synthesis protein NUDIX5; Nucleoside diphosphate-linked moiety X motif 5; nudix (nucleoside diphosphate linked moiety X)-type motif 5; Nudix motif 5; YSA1H
Gene Aliases: HSPC115; NUDIX5; NUDT5; YSA1; YSA1H; YSAH1
UniProt ID: (Human) Q9UKK9
Entrez Gene ID: (Human) 11164
Molecular Function: hydrolase pyrophosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.