R&D Systems
Manufacturer Code:AF6414SP
Catalog # AF6414SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human ADP-Sugar Pyrophosphatase/NUDT5 Glu2-Phe219 Accession # Q9UKK9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: 8-oxo-dGDP phosphatase; ADP-sugar pyrophosphatase; EC 3.6.1.- EC 3.6.1.13 hYSAH1 nucleoside diphosphate linked moiety X-type motif 5 Nucleoside diphosphate-linked moiety X motif 5 nudix (nucleoside diphosphate linked moiety X)-type motif 5 Nudix motif 5 YSA1 YSA1H YSA1HADP-sugar pyrophosphatase; Nuclear ATP-synthesis protein NUDIX5; Nucleoside diphosphate-linked moiety X motif 5; nudix (nucleoside diphosphate linked moiety X)-type motif 5; Nudix motif 5; YSA1H
Gene Aliases: HSPC115; NUDIX5; NUDT5; YSA1; YSA1H; YSAH1
UniProt ID: (Human) Q9UKK9
Entrez Gene ID: (Human) 11164
Molecular Function: hydrolase pyrophosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.