Invitrogen
Manufacturer Code:PA570463
Catalog # PIPA570463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human ADORA3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenosine receptor A3; ADORA3 A3AR AD026 Adenosine A3 receptor BA552M11.5 A3 adenosine receptor Adenosine receptor A3 RP11-552M11.7
Gene Aliases: A3AR; ADORA3
UniProt ID: (Bovine) Q0VC81, (Human) P0DMS8
Entrez Gene ID: (Pig) 606753, (Bovine) 780806, (Human) 140
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.