Invitrogen
Manufacturer Code:PA596008
Catalog # PIPA596008
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 100-200 of human ADORA2B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A2b, Rs; Adenosine receptor A2b
Gene Aliases: A2b; A2BAR; A2BR; AA2BR; Adora2b; AI480866; AI605384
UniProt ID: (Rat) P29276, (Mouse) Q60614
Entrez Gene ID: (Rat) 29316, (Mouse) 11541
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.