Invitrogen
Manufacturer Code:PA577849
Catalog # PIPA577849
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human Adenosine A2b Receptor. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A2b, Rs; Adenosine receptor A2b; ADORA2 ADORA2B adenosine A2b receptor
Gene Aliases: A2b; A2BAR; A2BR; AA2BR; ADORA2; ADORA2B; AI480866; AI605384
UniProt ID: (Human) P29275, (Rat) P29276, (Mouse) Q60614
Entrez Gene ID: (Human) 136, (Rat) 29316, (Mouse) 11541
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.