Invitrogen
Manufacturer Code:PA533321
Catalog # PIPA533321
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from 2nd extracellular domain of human Adenosine A1 Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenosine A1 receptor variant 1; adenosine A1 receptor variant 2; Adenosine receptor A1; ADORA1 A1 adenosine receptor A1ar Adenosine A1 receptor Adenosine receptor A1 ADO-A1 receptor RDC7
Gene Aliases: ADORA1; RDC7
UniProt ID: (Human) P30542, (Rat) P25099
Entrez Gene ID: (Human) 134, (Rat) 29290
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.