Invitrogen
Manufacturer Code:PA578733
Catalog # PIPA578733
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human ADO recombit protein (Position: E49-E261). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-aminoethanethiol (cysteamine) dioxygenase; 2-aminoethanethiol dioxygenase; 2-aminoethanethiol dioxygenase Cysteamine dioxygenase ADO C10orf22; cysteamine (2-aminoethanethiol) dioxygenase (ADO); Cysteamine dioxygenase
Gene Aliases: ADO; C10orf22; Gm237
UniProt ID: (Human) Q96SZ5, (Mouse) Q6PDY2
Entrez Gene ID: (Human) 84890, (Mouse) 211488
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.