Invitrogen
Manufacturer Code:PA552388
Catalog # PIPA552388
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ADNP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activity-dependent neuroprotector homeobox protein 2; ADNP homeobox protein 2; Zinc finger protein 508
Gene Aliases: 8430420L05Rik; ADNP2; BC024969; KIAA0863; mKIAA0863; RGD1308279; Zfp508; ZNF508
UniProt ID: (Human) Q6IQ32, (Mouse) Q8CHC8
Entrez Gene ID: (Human) 22850, (Rat) 307236, (Mouse) 240442
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.