Invitrogen
Manufacturer Code:PA599221
Catalog # PIPA599221
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ADNP corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA589558 activity dependent neuroprotector homeobox activity-dependent neuroprotective protein activity-dependent neuroprotector activity-dependent neuroprotector homeobox activity-dependent neuroprotector homeobox protein Adnp ADNP homeobox 1 ADNP1 HVDAS KIAA0784 mKIAA0784 MRD28 NAP peptide; Activity-dependent neuroprotective protein; activity-dependent neuroprotector; Activity-dependent neuroprotector homeobox protein; ADNP homeobox 1
Gene Aliases: AA589558; ADNP; ADNP1; HVDAS; KIAA0784; mKIAA0784; MRD28
UniProt ID: (Human) Q9H2P0, (Mouse) Q9Z103
Entrez Gene ID: (Human) 23394, (Mouse) 11538
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.