Novus Biologicals
Manufacturer Code:NBP297747AF700
Catalog # NB103234
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human ADNP fragment. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activity-dependent neuroprotective protein; Activity-dependent neuroprotective protein activity-dependent neuroprotector activity-dependent neuroprotector homeobox ADNP homeobox 1 ADNP1KIAA0784activity-dependent neuroprotector homeobox protein; activity-dependent neuroprotector; Activity-dependent neuroprotector homeobox protein; ADNP homeobox 1
Gene Aliases: ADNP; ADNP1; HVDAS; KIAA0784; MRD28
UniProt ID: (Human) Q9H2P0
Entrez Gene ID: (Human) 23394
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.