Proteintech
Manufacturer Code:237811AP150UL
Catalog # 50-173-4123
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ADM2 Fusion Protein Ag20548 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADM2; ADM2 adrenomedullin 2 AM2 IMDL Intermedin short IMDS Intermedin Intermedin long; Adrenomedullin-2; AM2; IMDL; IMDS; Intermedin; Intermedin-long; Intermedin-short; Protein ADM2
Gene Aliases: ADM2; AM2; dJ579N16.4; IMD; Imdn
UniProt ID: (Human) Q7Z4H4, (Mouse) Q7TNK8
Entrez Gene ID: (Human) 79924, (Mouse) 223780
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.