Novus Biologicals
Manufacturer Code:NBP197934AF532
Catalog # NP197934A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4F8 |
Immunogen | Recombinant human ADK (22-362aa) purified from E. coli |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Adenosine 5'-phosphotransferase; Adenosine kinase; adenosine kinase AKAdenosine 5&prime-phosphotransferase EC 2.7.1 EC 2.7.1.20; AK; testicular tissue protein Li 14
Gene Aliases: ADK; AK
UniProt ID: (Human) P55263
Entrez Gene ID: (Human) 132
Molecular Function:
carbohydrate kinase
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.