Abnova
Manufacturer Code:PAB19820
Catalog # 89-121-351
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human ADIPOR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ADIPOR1.
For Research Use Only
Protein Aliases: ACDCR1; Adiponectin receptor protein 1; CGI-45; CGI45; FLJ25385; FLJ42464; PAQR1; Progestin and adipoQ receptor family member 1; Progestin and adipoQ receptor family member I; TESBP1A
Gene Aliases: ACDCR1; ADIPOR1; CGI-45; CGI45; PAQR1; TESBP1A
UniProt ID: (Human) Q96A54
Entrez Gene ID: (Human) 51094
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.