Proteintech
Manufacturer Code:193591AP150UL
Catalog # 50-173-2388
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ADHFE1 Fusion Protein Ag6803 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADH8 ADHFE1 HMFT2263 HOT; ADHFe1; alcohol dehydrogenase 8; Alcohol dehydrogenase iron-containing protein 1; alcohol dehydrogenase, iron containing, 1; Fe-containing alcohol dehydrogenase; Fe-containing alcohol dehydrogenase 1; HOT; Hydroxyacid-oxoacid transhydrogenase, mitochondrial
Gene Aliases: 6330565B14Rik; ADH8; ADHFE1; AI043035; HMFT2263; HOT
UniProt ID: (Human) Q8IWW8, (Mouse) Q8R0N6, (Rat) Q4QQW3
Entrez Gene ID: (Human) 137872, (Mouse) 76187, (Rat) 362474
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.