Invitrogen
Manufacturer Code:PA542388
Catalog # PIPA542388
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ADH4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADH2; Alcohol dehydrogenase 4; Alcohol dehydrogenase 4 (class II) pi polypeptide ADH-2; alcohol dehydrogenase class II; Alcohol dehydrogenase class II pi chain; alcohol dehydrogenase II; aldehyde reductase; All-trans-retinol dehydrogenase [NAD(+)] ADH4; class II alcohol dehydrogenase, pi subunit; epididymis secretory protein Li 4
Gene Aliases: Ac1002; ADH-1; ADH-2; ADH4; HEL-S-4
UniProt ID: (Human) P08319
Entrez Gene ID: (Pig) 100513366, (Human) 127, (Bovine) 520508, (Dog) 478487, (Rat) 29646
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.