Abnova
Manufacturer Code:H00000127A01
Catalog # 89-003-214
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ADH4 (NP_000661 52 a.a. ~ 150 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant ADH4.
For Research Use Only
Protein Aliases: ADH-2; Alcohol dehydrogenase 4; Alcohol dehydrogenase class II pi chain; aldehyde reductase; All-trans-retinol dehydrogenase [NAD(+)] ADH4; epididymis secretory protein Li 4
Gene Aliases: ADH-2; ADH4; HEL-S-4
UniProt ID: (Human) P08319
Entrez Gene ID: (Human) 127
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.