Invitrogen
Manufacturer Code:PA5109363
Catalog # PIPA5109363
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 206-375 of human ADH1B (NP_000659.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2700007F14Rik ADH1B ADH2 Alcohol dehydrogenase 1B Alcohol dehydrogenase subunit beta Aldehyde dehydrogenase 5 Aldehyde dehydrogenase family 1 member B1 Aldehyde dehydrogenase X mitochondrial ALDH1B1 ALDH5 ALDHX All-trans-retinol dehydrogenase [NAD(+)] ADH1B HEL-S-117; acetaldehyde dehydrogenase 5; ADH, beta subunit; Alcohol dehydrogenase 1B; alcohol dehydrogenase 2 (class I), beta polypeptide; Alcohol dehydrogenase subunit beta; aldehyde dehydrogenase 1 family, member B1; aldehyde dehydrogenase 1B1; Aldehyde dehydrogenase 5; Aldehyde dehydrogenase family 1 member B1; Aldehyde dehydrogenase X, mitochondrial; aldehyde reductase; ALDH class 2; All-trans-retinol dehydrogenase [NAD(+)] ADH1B; epididymis secretory protein Li 117
Gene Aliases: ADH1B; ADH2; ALDH1B1; ALDH5; ALDHX; HEL-S-117
UniProt ID: (Human) P00325, (Human) P30837, (Rat) Q66HF8
Entrez Gene ID: (Human) 125, (Human) 219, (Rat) 298079
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.