Invitrogen
Manufacturer Code:PA536044
Catalog # PIPA536044
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 700-750 of human Adducin alpha |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Add97; adducin 1 (alpha); adducin 2 (beta); adducin beta; Adducin-63; alpha-ADD; Alpha-adducin; Alpha-adducin Erythrocyte adducin subunit alpha ADD1 ADDA alpha-Adducin Adducin1 Adducin-1 alpha Adducin Adducin-alpha Beta-adducin Erythrocyte adducin subunit beta ADD2 ADDB beta-Adducin Adducin1 Adducin-1 beta Adducin Adducin beta Adducin-beta; Beta-adducin; erythrocyte adducin alpha subunit; Erythrocyte adducin subunit alpha; Erythrocyte adducin subunit beta; snoRNA MBII-396
Gene Aliases: 2900072M03Rik; ADD1; ADD2; add97; ADDA; ADDB; AI256389; beta-ADD
UniProt ID: (Human) P35611, (Human) P35612, (Mouse) Q9QYC0, (Rat) Q63028, (Mouse) Q9QYB8, (Rat) Q05764
Entrez Gene ID: (Human) 118, (Human) 119, (Mouse) 11518, (Rat) 24170, (Mouse) 11519, (Rat) 24171
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.