Invitrogen
Manufacturer Code:PA530895
Catalog # PIPA530895
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 165 and 404 of Human Adenylate cyclase 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3',5'-cyclic AMP synthetase; AC2 HBAC2 3&prime5&prime-cyclic AMP synthetase ATP pyrophosphate-lyase 2 adenylate cyclase II adenylate cyclase type 2 adenylate cyclase type II adenylyl cyclase 2 type II adenylate cyclase; adenylate cyclase 2 (brain); adenylate cyclase II; Adenylate cyclase type 2; Adenylate cyclase type II; Adenylyl cyclase 2; ATP pyrophosphate-lyase 2; type II adenylate cyclase
Gene Aliases: AC2; ADCY2; HBAC2; KIAA1060
UniProt ID: (Human) Q08462
Entrez Gene ID: (Human) 108
Molecular Function: adenylate cyclase cyclase guanylate cyclase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.