Invitrogen
Manufacturer Code:PA5114701
Catalog # PIPA5114701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ADCY2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3',5'-cyclic AMP synthetase; 3'5'-cyclic AMP synthetase AC2 Adcy2 adenylate cyclase 2 adenylate cyclase 2 (brain) adenylate cyclase II adenylate cyclase type 2 Adenylate cyclase type II adenylyl cyclase 2 ATP pyrophosphate-lyase 2 HBAC2 KIAA1060 LOC100009201 mKIAA1060 type II adenylate cyclase type II adenylyl cyclase; AC2; adenylate cyclase 2 (brain); adenylate cyclase II; Adenylate cyclase type 2; Adenylate cyclase type II; Adenylyl cyclase 2; ATP pyrophosphate-lyase 2; type II adenylate cyclase
Gene Aliases: AC2; ADCY2; HBAC2; KIAA1060; mKIAA1060
UniProt ID: (Human) Q08462, (Rat) P26769
Entrez Gene ID: (Human) 108, (Mouse) 210044, (Rat) 81636
Molecular Function: adenylate cyclase cyclase guanylate cyclase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.