Abnova
Manufacturer Code:H00000108A01
Catalog # 89-104-042
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 977 a.a. to 1086 a.a) partial recombinant protein with GST tag.,ADCY2 (NP_065433 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant ADCY2.
For Research Use Only
Protein Aliases: 3',5'-cyclic AMP synthetase; AC2; adenylate cyclase 2 (brain); adenylate cyclase II; Adenylate cyclase type 2; Adenylate cyclase type II; Adenylyl cyclase 2; ATP pyrophosphate-lyase 2; FLJ16822; FLJ45092; HBAC2; KIAA1060; MGC133314; type II adenylate cyclase
Gene Aliases: AC2; ADCY2; HBAC2; KIAA1060
UniProt ID: (Human) Q08462
Entrez Gene ID: (Human) 108
Molecular Function: adenylate cyclase cyclase guanylate cyclase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.