Invitrogen
Manufacturer Code:PA513893
Catalog # PIPA513893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human ADCK5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AarF domain containing kinase 5 ADCK5; Uncharacterized aarF domain-containing protein kinase 5
Gene Aliases: A230108P17; ADCK5
UniProt ID: (Human) Q3MIX3, (Mouse) Q80V03
Entrez Gene ID: (Human) 203054, (Mouse) 268822
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.