Proteintech
Manufacturer Code:175791AP150UL
Catalog # 50-173-1662
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ADCK5 Fusion Protein Ag11094 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADCK5; Uncharacterized aarF domain-containing protein kinase 5
Gene Aliases: A230108P17; ADCK5
UniProt ID: (Human) Q3MIX3, (Mouse) Q80V03
Entrez Gene ID: (Human) 203054, (Rat) 362943, (Mouse) 268822
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.