Invitrogen
Manufacturer Code:PA555565
Catalog # PIPA555565
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ADCK4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aarF domain containing kinase 4; AarF domain-containing protein kinase 4; Atypical kinase COQ8B, mitochondrial; Coenzyme Q protein 8B; Uncharacterized aarF domain-containing protein kinase 4 aarF domain-containing protein kinase 4 ADCK4
Gene Aliases: ADCK4; COQ8B; NPHS9
UniProt ID: (Human) Q96D53
Entrez Gene ID: (Human) 79934
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.