Invitrogen
Manufacturer Code:PA5104233
Catalog # PIPA5104233
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ADCK1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610005A10Rik aarF domain containing kinase 1 AarF domain-containing protein kinase 1 Adck1 Nrbf1 uncharacterized aarF domain-containing protein kinase 1; AarF domain-containing protein kinase 1; uncharacterized aarF domain-containing protein kinase 1
Gene Aliases: 2610005A10Rik; ADCK1
UniProt ID: (Human) Q86TW2, (Mouse) Q9D0L4
Entrez Gene ID: (Human) 57143, (Rat) 366698, (Mouse) 72113
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.