Invitrogen
Manufacturer Code:PA578849
Catalog # PIPA578849
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human AZIN2 recombit protein (Position: C111-Q301). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADC; Antizyme inhibitor 2; Antizyme inhibitor 2 AzI2 Arginine decarboxylase ADC ARGDC Ornithine decarboxylase-like protein ODC-like protein ornithine decarboxylase paralog ODC-p AZIN2 ADC KIAA1945 ODCP; Arginine decarboxylase; Arginine decarboxylase-like protein; AzI2; ODC antizyme inhibitor-2; ODC-like protein; ODC-p; ODC-paralogue; ornithine decarboxylase like; ornithine decarboxylase paralog; Ornithine decarboxylase-like protein; ornithine decarboxylase-paralog
Gene Aliases: 4933429I20Rik; ADC; AZI2; AZIB1; AZIN2; B930082O19; KIAA1945; ODC-p; ODC1L; ODCP; RGD1564776
UniProt ID: (Human) Q96A70, (Mouse) Q8BVM4
Entrez Gene ID: (Human) 113451, (Mouse) 242669, (Rat) 366473
Molecular Function:
decarboxylase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.