Novus Biologicals
Manufacturer Code:NBP2843940.1ML
Catalog # NB032322
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human ADAT3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenosine deaminase tRNA-specific 3 TAD3 homolog (S. cerevisiae) FWP005 MST121 MSTP121 S863-5 TAD3 tRNA-specific adenosine deaminase 3 homolog tRNA-specific adenosine deaminase-like protein 3 tRNA-specific adenosine-34 deaminase subunit ADAT3; adenosine deaminase, tRNA-specific 3; adenosine deaminase, tRNA-specific 3, TAD3 homolog; Probable inactive tRNA-specific adenosine deaminase-like protein 3; tRNA-specific adenosine deaminase 3 homolog; tRNA-specific adenosine deaminase-like protein 3; tRNA-specific adenosine-34 deaminase subunit ADAT3
Gene Aliases: ADAT3; FWP005; MRT36; MST121; MSTP121; S863-5; TAD3
UniProt ID: (Human) Q96EY9
Entrez Gene ID: (Human) 113179
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.