Invitrogen
Manufacturer Code:PA570759
Catalog # PIPA570759
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human ADAT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenosine deaminase, tRNA-specific 2; adenosine deaminase, tRNA-specific 2, TAD2 homolog; deaminase domain containing 1; Deaminase domain-containing protein 1; tRNA-specific adenosine deaminase 2; tRNA-specific adenosine deaminase 2 homolog; tRNA-specific adenosine-34 deaminase subunit ADAT2
Gene Aliases: 4933426M09Rik; ADAT2; DEADC1; dJ20N2; dJ20N2.1; si:ch211-157l15.2; TAD2
UniProt ID: (Bovine) Q5E9J7, (Human) Q7Z6V5, (Mouse) Q6P6J0, (Zebrafish) Q5RIV4
Entrez Gene ID: (Bovine) 615418, (Human) 134637, (Dog) 100686906, (Horse) 100067148, (Rabbit) 100353433, (Mouse) 66757, (Rat) 361453, (Guinea pig) 100733583, (Zebrafish) 100034610
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.