Abnova
Manufacturer Code:PAB22686
Catalog # 89-124-394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human ADAT2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant ADAT2.
For Research Use Only
Protein Aliases: adenosine deaminase, tRNA-specific 2; adenosine deaminase, tRNA-specific 2, TAD2 homolog; DEADC1; deaminase domain containing 1; Deaminase domain-containing protein 1; dJ20N2; dJ20N2.1; DKFZp686L1118; FLJ44213; TAD2; tRNA-specific adenosine deaminase 2; tRNA-specific adenosine deaminase 2 homolog; tRNA-specific adenosine-34 deaminase subunit ADAT2
Gene Aliases: ADAT2; DEADC1; dJ20N2; dJ20N2.1; TAD2
UniProt ID: (Human) Q7Z6V5
Entrez Gene ID: (Human) 134637
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.