Abnova
Manufacturer Code:H00134637K
Catalog # 89-207-748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ADAT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ADAT2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: adenosine deaminase, tRNA-specific 2; adenosine deaminase, tRNA-specific 2, TAD2 homolog; DEADC1 DKFZp686L1118 FLJ44213 TAD2 dJ20N2 dJ20N2.1; deaminase domain containing 1; Deaminase domain-containing protein 1; tRNA-specific adenosine deaminase 2; tRNA-specific adenosine deaminase 2 homolog; tRNA-specific adenosine-34 deaminase subunit ADAT2
Gene Aliases: ADAT2; DEADC1; dJ20N2; dJ20N2.1; TAD2
UniProt ID: (Human) Q7Z6V5
Entrez Gene ID: (Human) 134637
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.