Invitrogen
Manufacturer Code:PA534828
Catalog # PIPA534828
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment contains a sequence corresponding to a region within amino acids 34 and 273 (P78563) of ADAR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADAR2 DRABA2 DRADA2 RED1 RED1 homolog RNA editase RNA editing deaminase 1 RNA-editing deaminase 1 RNA-editing enzyme 1 adenosine deaminase adenosine deaminase B1 homolog of rat RED1 double-stranded RNA-specific editase 1 dsRNA adenosine deaminase DRADA2; adenosine deaminase, RNA-specific, B1 (homolog of rat RED1); adenosine deaminase, RNA-specific, B1 (RED1 homolog rat); Double-stranded RNA-specific editase 1; dsRNA adenosine deaminase; dsRNA adenosine deaminase DRADA2; RED1 homolog; RNA editase; RNA editing deaminase 1; RNA-editing deaminase 1; RNA-editing enzyme 1
Gene Aliases: ADAR2; ADARB1; DRABA2; DRADA2; RED1
UniProt ID: (Human) P78563
Entrez Gene ID: (Human) 104
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.