Invitrogen
Manufacturer Code:PA5116693
Catalog # PIPA5116693
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 180-380 of human ADARB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700057H01Rik ADAR2 Adarb1 adenosine deaminase RNA specific B1 adenosine deaminase RNA-specific B1 adenosine deaminase RNA-specific B1 (homolog of rat RED1) adenosine deaminase RNA-specific B1 (RED1 homolog rat) AW124433 AW558573 BB220382 D10Bwg0447e Double-stranded RNA-specific editase 1 DRABA2 DRADA2 dsRNA adenosine deaminase dsRNA adenosine deaminase DRADA2 RED1 RED1 homolog RNA editase RNA editing deaminase 1 RNA editing deaminase of glutamate receptors RNA-editing deaminase 1 RNA-editing enzyme 1; adenosine deaminase, RNA-specific, B1 (homolog of rat RED1); adenosine deaminase, RNA-specific, B1 (RED1 homolog rat); Double-stranded RNA-specific editase 1; dsRNA adenosine deaminase; dsRNA adenosine deaminase DRADA2; RED1 homolog; RNA editase; RNA editing deaminase 1; RNA editing deaminase of glutamate receptors; RNA-editing deaminase 1; RNA-editing enzyme 1
Gene Aliases: 1700057H01Rik; ADAR2; ADARB1; AW124433; AW558573; BB220382; D10Bwg0447e; DRABA2; DRADA2; RED1
UniProt ID: (Human) P78563, (Rat) P51400, (Mouse) Q91ZS8
Entrez Gene ID: (Human) 104, (Rat) 25367, (Mouse) 110532
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.