Proteintech
Manufacturer Code:222481AP150UL
Catalog # 50-173-3795
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ADARB1 Fusion Protein Ag17743 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADAR2 ADAR2a ADAR2a L1 ADAR2a L2 ADAR2a L3 ADAR2b ADAR2c ADAR2d ADAR2g ADARB1 DRABA2 DRADA2 dsRNA adenosine deaminase RED1 RNA editing deaminase 1 RNA editing enzyme 1; adenosine deaminase, RNA-specific, B1 (homolog of rat RED1); adenosine deaminase, RNA-specific, B1 (RED1 homolog rat); Double-stranded RNA-specific editase 1; dsRNA adenosine deaminase; dsRNA adenosine deaminase DRADA2; RED1 homolog; RNA editase; RNA editing deaminase 1; RNA editing deaminase of glutamate receptors; RNA-editing deaminase 1; RNA-editing enzyme 1
Gene Aliases: ADAR2; ADARB1; DRABA2; DRADA2; RED1
UniProt ID: (Human) P78563, (Rat) P51400
Entrez Gene ID: (Human) 104, (Rat) 25367
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.