Proteintech
Manufacturer Code:153041AP150UL
Catalog # 50-173-0137
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ADAMTSL4 Fusion Protein Ag7781 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADAMTS like 4 ADAMTS like protein 4 ADAMTSL 4 ADAMTSL4 TSRC1 UNQ2803/PRO34012; ADAMTS-like protein 4; ADAMTSL-4; adamtsl4; adamtsl4 {ECO:0000312|EMBL:AAH99119.1}; thrombospondin repeat containing 1; thrombospondin repeat protein 1; Thrombospondin repeat-containing protein 1
Gene Aliases: ADAMTSL-4; ADAMTSL4; ECTOL2; PP1396; TSRC1; UNQ2803/PRO34012
UniProt ID: (Human) Q6UY14, (Rat) Q4FZU4, (Mouse) Q80T21
Entrez Gene ID: (Human) 54507, (Rat) 310670, (Mouse) 229595
Molecular Function:
enzyme modulator
extracellular matrix glycoprotein
extracellular matrix protein
hydrolase
metalloprotease
protease
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.