Abnova
Manufacturer Code:H00054507K
Catalog # 89-210-384
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ADAMTSL4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ADAMTSL4 peptide using ARM Technology.
For Research Use Only
Protein Aliases: ADAMTS-like protein 4; ADAMTSL-4; Thrombospondin repeat-containing protein 1; TSRC1
Gene Aliases: ADAMTSL-4; ADAMTSL4; ECTOL2; PP1396; TSRC1; UNQ2803/PRO34012
UniProt ID: (Human) Q6UY14
Entrez Gene ID: (Human) 54507
Molecular Function:
enzyme modulator
extracellular matrix glycoprotein
extracellular matrix protein
hydrolase
metalloprotease
protease
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.