Invitrogen
Manufacturer Code:PA560972
Catalog # PIPA560972
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ADAMTS7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a disintegrin and metalloprotease with thrombospondin motifs-7 preproprotein; A disintegrin and metalloproteinase with thrombospondin motifs 7; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 7; ADAM-TS 7; COMPase
Gene Aliases: ADAM-TS 7; ADAM-TS7; ADAMTS-7; ADAMTS7
UniProt ID: (Human) Q9UKP4
Entrez Gene ID: (Human) 11173
Molecular Function:
enzyme modulator
extracellular matrix glycoprotein
extracellular matrix protein
hydrolase
metalloprotease
protease
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.