Abnova
Manufacturer Code:PAB18820
Catalog # 89-120-379
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids at C-terminus of human ADAMTS7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ADAMTS7.
For Research Use Only
Protein Aliases: a disintegrin and metalloprotease with thrombospondin motifs-7 preproprotein; A disintegrin and metalloproteinase with thrombospondin motifs 7; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 7; ADAM-TS 7; ADAM-TS7; COMPase; DKFZp434H204
Gene Aliases: ADAM-TS 7; ADAM-TS7; ADAMTS-7; ADAMTS7
UniProt ID: (Human) Q9UKP4
Entrez Gene ID: (Human) 11173
Molecular Function:
enzyme modulator
extracellular matrix glycoprotein
extracellular matrix protein
hydrolase
metalloprotease
protease
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.