R&D Systems
Manufacturer Code:MAB2198SP
Catalog # MAB2198SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 362810 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human ADAMTS5 Ser262-Pro622 Accession # Q9UNA0 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A disintegrin and metalloproteinase with thrombospondin motifs 11; A disintegrin and metalloproteinase with thrombospondin motifs 11 a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif 5 (aggrecanase-2) ADAM metallopeptidase with thrombospondin type 1 motif 5 ADAM-TS 11 ADAM-TS 5 ADAMTS11 ADAMTS-11 ADAM-TS5 ADAMTS-5 ADMP2 ADMP-2 ADMP-2A disintegrin and metalloproteinase with thrombospondin motifs 5 Aggrecanase 2 Aggrecanase-2 disintegrin-like and metalloprotease with thrombospondin type 1 motif 510ADAMTS11FLJ36738 EC 3.4.24 EC 3.4.24.- EC 3.4.24.14 EC 3.4.24.82; A disintegrin and metalloproteinase with thrombospondin motifs 5; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase-2); ADAM-TS 11; ADAM-TS 5; ADMP-2; Aggrecanase-2
Gene Aliases: ADAM-TS 11; ADAM-TS 5; ADAM-TS5; ADAMTS-11; ADAMTS-5; ADAMTS11; ADAMTS5; ADMP-2; ADMP2
UniProt ID: (Human) Q9UNA0
Entrez Gene ID: (Human) 11096
Molecular Function: enzyme modulator extracellular matrix glycoprotein extracellular matrix protein hydrolase metalloprotease protease protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.