Invitrogen
Manufacturer Code:PA527165
Catalog # PIPA527165
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Human disintegrin and metalloproteinase with thrombospondin motifs 5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A disintegrin and metalloproteinase with thrombospondin motifs 11; A disintegrin and metalloproteinase with thrombospondin motifs 5; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase-2); ADAM-TS 11; ADAM-TS 11 ADAM-TS 5 ADAM-TS5 ADAMTS-11 ADAMTS-5 ADAMTS11 ADMP-2 A disintegrin and metalloproteinase with thrombospondin motifs 5 a disintegrin and metalloproteinase with thrombospondin motifs 11 a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif 5 (aggrecanase-2) aggrecanase-2; ADAM-TS 5; ADAMTS-5; ADMP-2; Aggrecanase-2; Implantin
Gene Aliases: 9530092O11Rik; ADAM-TS 11; ADAM-TS 5; ADAM-TS5; ADAMTS-11; ADAMTS-5; ADAMTS1; ADAMTS11; ADAMTS5; ADMP-2; ADMP2; AI481094; ASMP-2
UniProt ID: (Human) Q9UNA0, (Mouse) Q9R001
Entrez Gene ID: (Human) 11096, (Mouse) 23794
Molecular Function:
enzyme modulator
extracellular matrix glycoprotein
extracellular matrix protein
hydrolase
metalloprotease
protease
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.