OriGene
Manufacturer Code:TA805430
Catalog # 50-168-0695
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2G8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 296-577 of human ADAMTS4 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A disintegrin and metalloproteinase with thrombospondin motifs 4; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 4; ADAM metallopeptidase with thrombospondin type 1 motif, 4; ADAM-TS 4; ADAM-TS4; ADAMTS-2 ADAMTS-4 ADMP-1; ADMP-1; Aggrecanase-1
Gene Aliases: ADAMTS-2; ADAMTS-4; ADAMTS4; ADMP-1; KIAA0688; UNQ769/PRO1563
UniProt ID: (Human) O75173
Entrez Gene ID: (Human) 9507
Molecular Function:
enzyme modulator
extracellular matrix glycoprotein
extracellular matrix protein
hydrolase
metalloprotease
protease
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.